水杨酸 ≥99.5% (by titrimetric analysis) for biochemical research
供应商: TCI
|
Danger
|
Synonyms:
2-羟基苯甲酸
H1342-25G
H1342-5G
TCIAH1342-25GEA
0
CNY
TCIAH1342-25G
TCIAH1342-5G
水杨酸 ≥99.5% (by titrimetric analysis) for biochemical research
水杨酸
分子式:
(HO)C₆H₄CO₂H 分子量: 138.12 g/mol 沸点: 211°C 闪点: 157 存储条件: Ambient |
MDL编号:
MFCD00002439 CAS号: 69-72-7 EINECS: 200-712-3 Merck Index: 14,08332 |
|
产品技术规格
Appearance | White powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 99.5 % |
Melting point | 158.0 to 161.0 deg-C |
Protein impurity test (SDS-PAGE) | to pass test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...