3,4,5-三甲氧基苯甲酸甲酯 ≥99.0% (by GC)

供应商: TCI

Synonyms: 3,4,5-Trimethoxybenzoic acid methyl ester, Methyl 3,4,5-trimethoxybenzoate, Trimethylgallic Acid Methyl Ester

T1092-25G T1092-500G
TCIAT1092-25GEA 0 CNY
TCIAT1092-25G TCIAT1092-500G
3,4,5-三甲氧基苯甲酸甲酯 ≥99.0% (by GC)
3,4,5-三甲氧基苯甲酸甲酯
分子式: C₁₁H₁₄O₅
分子量: 226.23 g/mol
沸点: 185°C
MDL编号: MFCD00008431
CAS号: 1916-07-0
EINECS: 217-629-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=57fe3823ead719249f8daeae798b5a5d026fd5c4&vid=57fe3823ead719249f8daeae798b5a5d026fd5c4&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Melting point 82.0 to 85.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR