9-氯甲基蒽 ≥97.0% (by titrimetric analysis) for HPLC labeling
供应商: TCI
|
Danger
|
A5502-1G
A5502-5G
TCIAA5502-1GEA
0
CNY
TCIAA5502-1G
TCIAA5502-5G
9-氯甲基蒽 ≥97.0% (by titrimetric analysis) for HPLC labeling
9-氯甲基蒽
分子式:
C₁₅H₁₁Cl 分子量: 226.71 g/mol 沸点: 371 溶点: 142 存储条件: Ambient |
MDL编号:
MFCD00001263 CAS号: 24463-19-2 EINECS: 246-274-7 UN号: 3261 ADR: 8 II |
|
产品技术规格
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (Argentmetric Titration) | min. 97.0 % |
Melting point | 138.0 to 142.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...