Potassium(2-methylphenyl)trifluoroborate ≥98.0% (by HPLC)
供应商: TCI
|
Warning
|
Synonyms:
(2-Methylphenyl)trifluoroboric acid potassium salt, Potassium trifluoro(o-tolyl)borate
P1818-1G
P1818-5G
TCIAP1818-1GEA
0
CNY
TCIAP1818-1G
TCIAP1818-5G
Potassium(2-methylphenyl)trifluoroborate ≥98.0% (by HPLC)
Potassium(2-methylphenyl)trifluoroborate
分子式:
C₇H₇BF₃K 分子量: 198.04 g/mol |
MDL编号:
MFCD03790006 CAS号: 274257-34-0 |
|
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Melting point | 224.0 to 228.0 deg-C |
Elemental analysis (Carbon) | 41.00 to 43.00 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...