1-碘癸烷 ≥96.0% (by GC) stabilised

供应商: TCI
Warning

Synonyms: 癸基碘

Stabilization: Stabilised with copper chip

I0494-25G I0494-250G
TCIAI0494-25GEA 0 CNY
TCIAI0494-25G TCIAI0494-250G
1-碘癸烷 ≥96.0% (by GC) stabilised
1-碘癸烷
分子式: C₁₀H₂₁I
分子量: 268.18 g/mol
沸点: 132°C
溶点: -16.3
存储条件: Ambient
MDL编号: MFCD00001085
CAS号: 2050-77-3
EINECS: 218-104-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c5da403a20df291949a8e190f416d7eddc593369&vid=c1569cf257611bf62233757fbb98dff45336ab22&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light yellow to Light red clear liquid
Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR