全氟丁酰氯 ≥95.0% (by GC) for heptafluorobutyration

供应商: TCI
Danger

Synonyms: 七氟丁酰氯

H0508-25G H0508-5G
TCIAH0508-25GEA 0 CNY
TCIAH0508-25G TCIAH0508-5G
全氟丁酰氯 ≥95.0% (by GC) for heptafluorobutyration
全氟丁酰氯
分子式: C₄ClF₇O
分子量: 232.48 g/mol
沸点: 39°C
存储条件: Refrigerator
MDL编号: MFCD00000707
CAS号: 375-16-6
EINECS: 206-785-8
UN号: 3265
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b5a6a73240ca4528cbda7cfc5eee8037b76ef018&vid=cfec4481e8f1f886f7b38c0f83db98ecc6647056&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR