2,3-Difluoro-6-methoxybenzaldehyde ≥97.0% (by GC)

供应商: TCI
Warning

D3885-1G D3885-5G
TCIAD3885-1GEA 0 CNY
TCIAD3885-1G TCIAD3885-5G
2,3-Difluoro-6-methoxybenzaldehyde ≥97.0% (by GC)
2,3-Difluoro-6-methoxybenzaldehyde
分子式: C₈H₆F₂O₂
MDL编号: MFCD02093968
CAS号: 187543-87-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aec940e3bf1a058ce29f723d31da4f016230ac2d&vid=aec940e3bf1a058ce29f723d31da4f016230ac2d&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 97.0 %
Melting point 56.0 to 60.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR