全氟己基溴 ≥97.0% (by GC)

供应商: TCI
Warning

Synonyms: 1-溴全氟己烷

T2479-25G T2479-5G
TCIAT2479-25GEA 0 CNY
TCIAT2479-25G TCIAT2479-5G
全氟己基溴 ≥97.0% (by GC)
全氟己基溴
分子式: BrF₂C(CF₂)₄CF₃
分子量: 398.95 g/mol
沸点: 100°C
MDL编号: MFCD00042349
CAS号: 335-56-8
EINECS: 206-391-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4f3643834454fee379b49ee49653d24490f7d75d&vid=2d8e63a42bce1457756266bd7b18eeb316a81b57&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR