2-甲基环己醇,顺反异构体混合物 cis- and trans-mixture ≥98.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
六氢邻甲酚
M0194-25ML
M0194-500ML
TCIAM0194-25MLEA
0
CNY
TCIAM0194-25ML
TCIAM0194-500ML
2-甲基环己醇,顺反异构体混合物 cis- and trans-mixture ≥98.0% (by GC)
2-甲基环己醇,顺反异构体混合物
分子式:
C₇H₁₄O 分子量: 114.19 g/mol 沸点: 163 溶点: -38 闪点: 58°C 存储条件: Ambient |
MDL编号:
MFCD00001443 CAS号: 583-59-5 EINECS: 209-512-0 UN号: 2617 ADR: 3 III |
|
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % (total of isomers) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...