3-噻吩硼酸 (contains varying amounts of Anhydride)
供应商: TCI
|
Warning
|
T1975-1G
T1975-5G
TCIAT1975-1GEA
0
CNY
TCIAT1975-1G
TCIAT1975-5G
3-噻吩硼酸 (contains varying amounts of Anhydride)
3-噻吩硼酸
分子式:
C₄H₅BO₂S 溶点: 162…164 °C |
MDL编号:
MFCD00151851 CAS号: 6165-69-1 |
|
产品技术规格
Appearance | White to Light yellow powder to crystal |
Purity (Neutralization titration) | 97.0 and le 116.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...