2-氯吡啶-3-硼酸 (contains varying amounts of Anhydride)
供应商: TCI
|
Warning
|
Synonyms:
2-Chloro-3-pyridinylboronic acid, (2-Chloropyridin-3-yl)boronic acid, 2-Chloropyridine-3-boronic acid
C2560-1G
C2560-5G
TCIAC2560-1GEA
0
CNY
TCIAC2560-1G
TCIAC2560-5G
2-氯吡啶-3-硼酸 (contains varying amounts of Anhydride)
2-氯吡啶-3-硼酸
分子式:
C₅H₅BClNO₂ 分子量: 157.36 g/mol 溶点: 160°C |
MDL编号:
MFCD03094997 CAS号: 381248-04-0 |
|
产品技术规格
Appearance | White to Light yellow powder to crystal |
Purity (Neutralization titration) | 97.0 to 113.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...