3-乙酰苯基硼酸 (contains varying amounts of Anhydride)

供应商: TCI
Warning

Synonyms: 3'-Boronoacetophenone, 3-Acetylphenylboronic Acid, 3-Acetylbenzeneboronic acid, Dihydroxy(3-acetylphenyl)borane, (3-Acetylphenyl)metaboric acid, 3-Acetylphenylboronic acid, 3-Acetylbenzeneboronic acid

A1942-25G A1942-5G
TCIAA1942-25GEA 0 CNY
TCIAA1942-25G TCIAA1942-5G
3-乙酰苯基硼酸 (contains varying amounts of Anhydride)
3-乙酰苯基硼酸
分子式: C₈H₉BO₃
MDL编号: MFCD01074678
CAS号: 204841-19-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2493495e2c0703a7110e170b296b8d20a8d12f9d&vid=2493495e2c0703a7110e170b296b8d20a8d12f9d&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Light orange to Yellow to Green powder to crystal
Purity (Neutralization titration) 95.0 to 112.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR