3-(溴甲基)苯硼酸 (contains varying amounts of Anhydride)

供应商: TCI
Danger

B3814-1G B3814-5G
TCIAB3814-1GEA 0 CNY
TCIAB3814-1G TCIAB3814-5G
3-(溴甲基)苯硼酸 (contains varying amounts of Anhydride)
3-(溴甲基)苯硼酸
分子式: C₇H₈BBrO₂
溶点: 216°C
存储条件: Refrigerator
MDL编号: MFCD01632207
CAS号: 51323-43-4
UN号: 3261
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=62df46f129b539b03115dd38ad5df9ce54ad54ce&vid=62df46f129b539b03115dd38ad5df9ce54ad54ce&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Argentmetric Titration) 97.0 to 110.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR