1-乙基-3-甲基咪唑六氟磷酸盐 ≥98.0% (by titrimetric analysis) for molten salt

供应商: TCI
Warning

Synonyms: EMIMPF6

E0493-25G E0493-5G
TCIAE0493-25GEA 0 CNY
TCIAE0493-25G TCIAE0493-5G
1-乙基-3-甲基咪唑六氟磷酸盐 ≥98.0% (by titrimetric analysis) for molten salt
1-乙基-3-甲基咪唑六氟磷酸盐
分子式: C₆H₁₁F₆N₂P
分子量: 256.13 g/mol
MDL编号: MFCD00216703
CAS号: 155371-19-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b55cb29da5f67f6891d0428020348cf9fe5b8a21&vid=77f9ed30cb605bd5bd9436a4d20c143eb545de9a&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystaline
Purity (Ion exchange titration) min. 98.0 %
Melting point 59.0 to 62.0 deg-C
Solubility in hot Water very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR