3-溴苯乙醇 ≥98.0% (by GC)

供应商: TCI

B3517-25G B3517-5G
TCIAB3517-25GEA 0 CNY
TCIAB3517-25G TCIAB3517-5G
3-溴苯乙醇 ≥98.0% (by GC)
3-溴苯乙醇
分子式: C₈H₉BrO
分子量: 201.06 g/mol
沸点: 110°C
MDL编号: MFCD00191857
CAS号: 28229-69-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=17a19ce498384dbb6cb07c9de5523e7b456876b8&vid=17a19ce498384dbb6cb07c9de5523e7b456876b8&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR