1-(三甲基硅基)咪唑 ≥98.0% (by titrimetric analysis) trimethylsilylating reagent
供应商: TCI
|
Danger
|
Synonyms:
TMSI, N-三甲基硅基咪唑
T0585-100G
T0585-25G
TCIAT0585-100GEA
0
CNY
TCIAT0585-100G
TCIAT0585-25G
1-(三甲基硅基)咪唑 ≥98.0% (by titrimetric analysis) trimethylsilylating reagent
1-(三甲基硅基)咪唑
分子式:
C₆H₁₂N₂Si 沸点: 91°C 溶点: –42 °C 密度: 0,95 g/cm³ (20 °C) 闪点: 5°C 存储条件: Refrigerator |
MDL编号:
MFCD00005280 CAS号: 18156-74-6 EINECS: 242-040-3 UN号: 1993 ADR: 3,II |
|
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (Nonaqueous Titration) | min. 98.0 % |
Effect test of GC derivalization | Effective as derivatization agent D- (+)-Glucose |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...