4-Phenylbenzyl bromide ≥98.0% (by GC)

供应商: TCI
Warning

Synonyms: 4-(Bromomethyl)biphenyl, 4-(Bromomethyl)-1,1'-biphenyl, p-(Bromomethyl)biphenyl

B1847-25G B1847-5G
TCIAB1847-25GEA 0 CNY
TCIAB1847-25G TCIAB1847-5G
4-Phenylbenzyl bromide ≥98.0% (by GC)
4-Phenylbenzyl bromide
分子式: C₁₃H₁₁Br
分子量: 247.13 g/mol
沸点: 140°C
MDL编号: MFCD00017869
CAS号: 2567-29-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a24c01b09b2359c85ec89066751ca36e4ec961ae&vid=a24c01b09b2359c85ec89066751ca36e4ec961ae&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 84.0 to 87.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR