3-(溴甲基)苯甲腈 ≥98.0% (by GC)

供应商: TCI
Danger

Synonyms: α-溴间甲基苯甲腈, 3-氰基溴苄

C1665-25G C1665-5G
TCIAC1665-25GEA 0 CNY
TCIAC1665-25G TCIAC1665-5G
3-(溴甲基)苯甲腈 ≥98.0% (by GC)
3-(溴甲基)苯甲腈
分子式: C₈H₆BrN
分子量: 196.05 g/mol
沸点: 130°C
MDL编号: MFCD00001809
CAS号: 28188-41-2
EINECS: 248-890-1
UN号: 3261
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9db5ca27c615c1daaa22ae8d25a066d6fc803ed5&vid=9db5ca27c615c1daaa22ae8d25a066d6fc803ed5&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystaline
Purity (GC) min. 98.0 %
Melting point 93.0 to 96.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR