(2-Nitrophenoxy)acetic acid ≥98.0% (by GC, titration analysis)

供应商: TCI
Warning

Synonyms: 2-(2-Nitrophenoxy)acetic acid

N0225-25G N0225-5G
TCIAN0225-25GEA 0 CNY
TCIAN0225-25G TCIAN0225-5G
(2-Nitrophenoxy)acetic acid ≥98.0% (by GC, titration analysis)
(2-Nitrophenoxy)acetic acid
分子式: (C₆H₄NO₂)OCH₂COOH
MDL编号: MFCD00016993
CAS号: 1878-87-1
EINECS: 217-527-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6b2053a483a9f98e8eb8faffceade7f93ed41630&vid=6b2053a483a9f98e8eb8faffceade7f93ed41630&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 156.0 to 160.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR