L-(-)-乳酸甲酯 ≥98.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
(S)-(-)-乳酸甲酯
L0163-25G
L0163-500G
TCIAL0163-25GEA
0
CNY
TCIAL0163-25G
TCIAL0163-500G
L-(-)-乳酸甲酯 ≥98.0% (by GC)
L-(-)-乳酸甲酯
分子式:
C₄H₈O₃ 分子量: 104.11 g/mol 沸点: 140°C 溶点: -44 闪点: 49°C |
MDL编号:
MFCD00064265 CAS号: 27871-49-4 EINECS: 248-704-9 UN号: 3272 ADR: 3 III |
|
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Optical purity (GC) | min. 97.0 ee% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...