2-Fluoromesitylene ≥98.0% (by GC)

供应商: TCI
Warning

Synonyms: 2-Fluoro-1,3,5-trimethylbenzene, Fluoromesitylene, 2,4,6-Trimethylfluorobenzene

M1201-1G M1201-5G
TCIAM1201-1GEA 0 CNY
TCIAM1201-1G TCIAM1201-5G
2-Fluoromesitylene ≥98.0% (by GC)
2-Fluoromesitylene
分子式: C₉H₁₁F
分子量: 138.18 g/mol
沸点: 165°C
MDL编号: MFCD00052365
CAS号: 392-69-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9eb1f31ccc58e4047a949e7af5217c2df4ab2baa&vid=9eb1f31ccc58e4047a949e7af5217c2df4ab2baa&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR