二丁醚 ≥99.0% (by GC) stabilised

供应商: TCI
Danger

Synonyms: 丁醚, DBE

Stabilization: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)

B0721-25ML B0721-500ML
TCIAB0721-25MLEA 0 CNY
TCIAB0721-25ML TCIAB0721-500ML
二丁醚 ≥99.0% (by GC) stabilised
二丁醚
分子式: C₈H₁₈O
分子量: 130.23 g/mol
沸点: 141°C
溶点: -95
闪点: 37°C
存储条件: Refrigerator
MDL编号: MFCD00009461
CAS号: 142-96-1
EINECS: 205-575-3
UN号: 1149
ADR: 3 III
Merck Index: 14,01569
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b9301fcbf7c34792d7845c5193272fd4452e9c40&vid=b9301fcbf7c34792d7845c5193272fd4452e9c40&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless clear liquid
Purity (GC) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR