D-(-)-酒石酸二甲酯 ≥98.0% (by GC)
供应商: TCI
Synonyms:
D-(-)-Tartaric acid dimethyl ester, 二甲基 D-酒石酸酯
T1659-25G
T1659-5G
TCIAT1659-25GEA
0
CNY
TCIAT1659-25G
TCIAT1659-5G
D-(-)-酒石酸二甲酯 ≥98.0% (by GC)
D-(-)-酒石酸二甲酯
分子式:
C₆H₁₀O₆ 分子量: 178.14 g/mol 沸点: 118°C |
MDL编号:
MFCD00008445 CAS号: 13171-64-7 EINECS: 236-118-6 |
|
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 60.0 to 65.0 deg-C |
Specific rotation [a]20/D | -20.0 to -22.0 deg (C=1, H2O) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...