二丁基硫醚 ≥98.0% (通过GC)

供应商: TCI

B0740-25ML B0740-500ML
TCIAB0740-25MLEA 0 CNY
TCIAB0740-25ML TCIAB0740-500ML
二丁基硫醚 ≥98.0% (通过GC)
二丁基硫醚
分子式: C₈H₁₈S
沸点: 189 °C (1013 hPa)
溶点: –80 °C
密度: 0,83 g/cm³ (20 °C)
闪点: 76°C
MDL编号: MFCD00009468
CAS号: 544-40-1
EINECS: 208-870-5
ADR: 9,
Merck Index: 14,01590
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=192c9a91fb2dda4423d6e8e29325975b39103e85&vid=192c9a91fb2dda4423d6e8e29325975b39103e85&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR