2,3,4,5,6-五氟苯甲醇 ≥96.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
(Perfluorophenyl)methanol, (2,3,4,5,6-Pentafluorophenyl)methanol
P0849-25G
P0849-5G
TCIAP0849-25GEA
0
CNY
TCIAP0849-25G
TCIAP0849-5G
2,3,4,5,6-五氟苯甲醇 ≥96.0% (by GC)
2,3,4,5,6-五氟苯甲醇
分子式:
C₇H₃F₅O 沸点: 182°C 溶点: 30…32 °C 存储条件: Ambient |
MDL编号:
MFCD00004602 CAS号: 440-60-8 EINECS: 207-126-7 |
|
产品技术规格
Appearance | White or Colorless to Light yellow powder to lump to clear liquid |
Purity (GC) | min. 96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...