六氟戊二酸酐 ≥97.0% (by titrimetric analysis)

供应商: TCI
Warning

Synonyms: 3,3,4,4,5,5-Hexafluorodihydro-2H-pyran-2,6(3H)-dione

H0745-25G H0745-5G
TCIAH0745-25GEA 0 CNY
TCIAH0745-25G TCIAH0745-5G
六氟戊二酸酐 ≥97.0% (by titrimetric analysis)
六氟戊二酸酐
分子式: C₅F₆O₃
沸点: 72°C
密度: 1,65
MDL编号: MFCD00006680
CAS号: 376-68-1
EINECS: 206-811-8
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f18321cb0f8944d817ae86b487e3e8730cd44d9&vid=2f18321cb0f8944d817ae86b487e3e8730cd44d9&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (Morpholine Method) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR