邻乙氧基苯硼酸 (contains varying amounts of Anhydride)
供应商: TCI
|
Warning
|
E0723-1G
E0723-5G
TCIAE0723-1GEA
0
CNY
TCIAE0723-1G
TCIAE0723-5G
邻乙氧基苯硼酸 (contains varying amounts of Anhydride)
邻乙氧基苯硼酸
分子式:
C₈H₁₁BO₃ 溶点: 103°C |
MDL编号:
MFCD00674027 CAS号: 213211-69-9 |
|
产品技术规格
Appearance | White to Light yellow powder to crystal |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...