2,4-二甲基-3-乙基吡咯 96%
供应商: Thermo Fisher Scientific
|
Warning
|
116210050
116210250
ACRO116210050EA
0
CNY
ACRO116210050
ACRO116210250
2,4-二甲基-3-乙基吡咯 96%
2,4-二甲基-3-乙基吡咯
分子式:
C₈H₁₃N 沸点: 197 °C (710 mmHg) 密度: 0,913 g/cm³ 闪点: 71 °C 存储条件: Refrigerator |
MDL编号:
MFCD00005222 CAS号: 517-22-6 EINECS: 208-234-7 |
产品技术规格
Appearance | Clear yellow to red-brown Liquid (May darken on storage) |
Infrared spectrum | Conforms |
Refractive index | 1.4960 to 1.4980 (20°C, 589 nm) |
GC | ≥95 % |
Titration excess NaOH | ≥97.0 % |
Chloride (Cl) | ≤5 ppm |
Nitrate (NO3) | ≤10 ppm |
Sulfate (SO4) | ≤10 ppm |
Iron (Fe) | ≤5 ppm |
Heavy metals (as Pb) | ≤5 ppm |
Insoluble matter | ≤0.005 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...