5-氮杂胞嘧啶 ≥98.0% (by titrimetric analysis)

供应商: TCI

Synonyms: 4-氨基-1,3,5-三嗪-2-酮

A1091-25G A1091-5G
TCIAA1091-25GEA 0 CNY
TCIAA1091-25G TCIAA1091-5G
5-氮杂胞嘧啶 ≥98.0% (by titrimetric analysis)
5-氮杂胞嘧啶
分子式: C₃H₄N₄O
分子量: 112.09 g/mol
MDL编号: MFCD00006033
CAS号: 931-86-2
EINECS: 213-242-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e73a286c9ec4a137cd6d0b31dea2091088256acd&vid=e73a286c9ec4a137cd6d0b31dea2091088256acd&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 % (calcd.on anh.substance)
Water max. 5.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR