1-苯基-1-环戊烷羧酸 ≥98.0% (by GC, titration analysis)

供应商: TCI

Synonyms: 1-Phenyl-1-cyclopentanecarboxylic acid

P1487-25G P1487-5G
TCIAP1487-25GEA 0 CNY
TCIAP1487-25G TCIAP1487-5G
1-苯基-1-环戊烷羧酸 ≥98.0% (by GC, titration analysis)
1-苯基-1-环戊烷羧酸
分子式: C₁₂H₁₄O₂
分子量: 190.24 g/mol
MDL编号: MFCD00001372
CAS号: 77-55-4
EINECS: 201-037-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5cac2c0ccd1e11229ea2fdf077ae6a4e25a80cf2&vid=33c92ad48f1090429520892c6a7cf09830a08504&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 157.0 to 161.0 deg-C
Solubility in Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR