1-苯基-1-环戊烷羧酸 98%

供应商: Thermo Fisher Scientific
Warning

Synonyms: 1-Phenyl-1-cyclopentanecarboxylic acid

161140250 161140050
ACRO161140250EA 0 CNY
ACRO161140250 ACRO161140050
1-苯基-1-环戊烷羧酸 98%
1-苯基-1-环戊烷羧酸
分子式: C₁₂H₁₄O₂
溶点: 157…161 °C
存储条件: Ambient
MDL编号: MFCD00001372
CAS号: 77-55-4
EINECS: 201-037-7

现在订购

产品技术规格

Appearance Cream to grey-brown Powder
Infrared spectrum Conforms
Melting point 157 to 161 °C
GC ≥97.5 %
Titration with NaOH ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR