4′-(甲硫基)苯乙酮 ≥98.0% (by GC)

供应商: TCI

Synonyms: 1-(4-(Methylthio)phenyl)ethanone

M1672-25G M1672-5G
TCIAM1672-25GEA 0 CNY
TCIAM1672-25G TCIAM1672-5G
4′-(甲硫基)苯乙酮 ≥98.0% (by GC)
4′-(甲硫基)苯乙酮
分子式: C₉H₁₀OS
分子量: 166.24 g/mol
MDL编号: MFCD00039835
CAS号: 1778-09-2
EINECS: 217-213-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=31b5ae8b2b0a243a3074ad9589e3fdb37ca5dd72&vid=31b5ae8b2b0a243a3074ad9589e3fdb37ca5dd72&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 79.0 to 84.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR