辛二酸双(N-羟基琥珀酰亚胺酯) ≥98.0% (by HPLC, total nitrogen)
供应商: TCI
Synonyms:
双琥珀酰亚胺辛二酸酯, 辛二酸二琥珀酰亚胺酯 (DSS)
D3895-1G
D3895-5G
TCIAD3895-1GEA
0
CNY
TCIAD3895-1G
TCIAD3895-5G
辛二酸双(N-羟基琥珀酰亚胺酯) ≥98.0% (by HPLC, total nitrogen)
辛二酸双(N-羟基琥珀酰亚胺酯)
分子式:
C₁₆H₂₀N₂O₈ 分子量: 368.34 g/mol |
MDL编号:
MFCD00049059 CAS号: 68528-80-3 |
|
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 169.0 to 173.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...