4-Aminocyclohexanecarboxylic acid (mixture of cis and trans isomers) cis- and trans-mixture ≥95.0% (by GC, titration analysis)

供应商: TCI

Synonyms: 4-Aminocyclohexanecarboxylic acid, 4-Aminocyclohexane-1-carboxylic acid

A1278-25G A1278-5G
TCIAA1278-25GEA 0 CNY
TCIAA1278-25G TCIAA1278-5G
4-Aminocyclohexanecarboxylic acid (mixture of cis and trans isomers) cis- and trans-mixture ≥95.0% (by GC, titration analysis)
4-Aminocyclohexanecarboxylic acid (mixture of cis and trans isomers)
分子式: C₇H₁₃NO₂
分子量: 143.1800 g/mol
MDL编号: MFCD00191601
CAS号: 1776-53-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=174bc572b634c1adb090a79c1902b5356ebd2368&vid=229cdc212949f86f15efafe9e2ccfa4709652539&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 95.0 % (total of isomers)
Purity (Nonaqueous Titration) min. 95.0 %
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR