1-(3-Aminopropyl)pyrrolidine ≥98.0% (通过滴定分析)

供应商: TCI
Danger

A2270-25G A2270-5G
TCIAA2270-25GEA 0 CNY
TCIAA2270-25G TCIAA2270-5G
1-(3-Aminopropyl)pyrrolidine ≥98.0% (通过滴定分析)
1-(3-Aminopropyl)pyrrolidine
分子式: C₇H₁₆N₂
沸点: 82°C
MDL编号: MFCD00014100
CAS号: 23159-07-1
EINECS: 245-465-2
UN号: 2735
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e8090f28a6d25d7745460b4d0285f9c3d2090528&vid=e8090f28a6d25d7745460b4d0285f9c3d2090528&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR