1-(2-氨乙基)吡咯烷 ≥98.0% (by GC, titration analysis)
供应商: TCI
|
Danger
|
Synonyms:
N-(2-氨基乙基)吡咯烷
A0884-25G
A0884-5G
TCIAA0884-25GEA
0
CNY
TCIAA0884-25G
TCIAA0884-5G
1-(2-氨乙基)吡咯烷 ≥98.0% (by GC, titration analysis)
1-(2-氨乙基)吡咯烷
分子式:
C₆H₁₄N₂ 分子量: 114.19 g/mol 沸点: 161°C 闪点: 47°C |
MDL编号:
MFCD00003182 CAS号: 7154-73-6 EINECS: 230-509-5 UN号: 2733 ADR: 3 III |
|
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...