trans-4-Propylcyclohexanecarboxylic acid ≥98.0% (by GC, titration analysis)
供应商: TCI
|
Warning
|
P0943-25G
P0943-5G
TCIAP0943-25GEA
0
CNY
TCIAP0943-25G
TCIAP0943-5G
trans-4-Propylcyclohexanecarboxylic acid ≥98.0% (by GC, titration analysis)
trans-4-Propylcyclohexanecarboxylic acid
分子式:
C₁₀H₁₈O₂ 分子量: 170.24 g/mol |
MDL编号:
MFCD00059559 CAS号: 38289-27-9 |
|
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Solubility in Methanol | almost transparency |
Melting point | 97.0 to 100.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...