trans-4-Ethylcyclohexanecarboxylic acid ≥98.0% (by GC, titration analysis)
供应商: TCI
|
Warning
|
E0904-25G
E0904-5G
TCIAE0904-25GEA
0
CNY
TCIAE0904-25G
TCIAE0904-5G
trans-4-Ethylcyclohexanecarboxylic acid ≥98.0% (by GC, titration analysis)
trans-4-Ethylcyclohexanecarboxylic acid
分子式:
C₉H₁₆O₂ 分子量: 156.23 g/mol |
MDL编号:
MFCD01076323 CAS号: 6833-47-2 |
|
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 50.0 to 54.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...