trans-4-Aminocyclohexanecarboxylic acid ≥98.0% (by GC, titration analysis)
供应商: TCI
A1656-1G
A1656-25G
A1656-5G
TCIAA1656-1GEA
0
CNY
TCIAA1656-1G
TCIAA1656-25G
TCIAA1656-5G
trans-4-Aminocyclohexanecarboxylic acid ≥98.0% (by GC, titration analysis)
trans-4-Aminocyclohexanecarboxylic acid
分子式:
C₇H₁₃NO₂ 分子量: 143.19 g/mol 溶点: 495°C |
MDL编号:
MFCD00209953 CAS号: 3685-25-4 |
|
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...