2-Methyltryptamine ≥98.0% (by GC, titration analysis)

供应商: TCI
Warning

Synonyms: 3-(2-Aminoethyl)-2-methylindole

M2055-1G M2055-5G
TCIAM2055-1GEA 0 CNY
TCIAM2055-1G TCIAM2055-5G
2-Methyltryptamine ≥98.0% (by GC, titration analysis)
2-Methyltryptamine
分子式: C₁₁H₁₄N₂
沸点: 177°C
存储条件: Refrigerator
MDL编号: MFCD00130185
CAS号: 2731-06-8
EINECS: 220-347-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5a64d64f32f39d3038bb9990bedf11137a88a39e&vid=5a64d64f32f39d3038bb9990bedf11137a88a39e&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow to Dark green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 106.0 to 110.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR