Decyl decanoate ≥95.0% (by GC)
供应商: TCI
D1080-25ML
D1080-500ML
TCIAD1080-25MLEA
0
CNY
TCIAD1080-25ML
TCIAD1080-500ML
Decyl decanoate ≥95.0% (by GC)
Decyl decanoate
分子式:
C₂₀H₄₀O₂ 分子量: 312.5304 g/mol 沸点: 159°C 闪点: 140°C |
MDL编号:
MFCD00059280 CAS号: 1654-86-0 EINECS: 216-729-4 |
|
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...