2,5-Dichloroterephthalic acid ≥97.0% (通过滴定分析)

供应商: TCI
Warning

D1698-1G D1698-5G
TCIAD1698-1GEA 0 CNY
TCIAD1698-1G TCIAD1698-5G
2,5-Dichloroterephthalic acid ≥97.0% (通过滴定分析)
2,5-Dichloroterephthalic acid
分子式: C₈H₄Cl₂O₄
MDL编号: MFCD00058982
CAS号: 13799-90-1
EINECS: 237-454-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a3a2fc8400e0d661cfdb26687301e7ac59ed756c&vid=a3a2fc8400e0d661cfdb26687301e7ac59ed756c&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 97.0 %
Solubility in Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR