2,6-二苯基苯酚 ≥98.0% (by GC)

供应商: TCI
Warning

Synonyms: [1,1':3',1''-Terphenyl]-2'-ol

D2563-25G D2563-5G
TCIAD2563-25GEA 0 CNY
TCIAD2563-25G TCIAD2563-5G
2,6-二苯基苯酚 ≥98.0% (by GC)
2,6-二苯基苯酚
分子式: C₁₈H₁₄O
MDL编号: MFCD00009716
CAS号: 2432-11-3
EINECS: 219-401-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a31194a03fe17152fb743915a3344c7d8135819e&vid=a31194a03fe17152fb743915a3344c7d8135819e&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 101.0 to 104.0 deg-C
Solubility in Toluene almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR