D-叔亮氨酸 ≥98.0% (by titrimetric analysis)

供应商: TCI

Synonyms: (R)-2-氨基-3,3-二甲基丁酸, D-α-叔丁基甘氨酸

L0157-1G L0157-5G
TCIAL0157-1GEA 0 CNY
TCIAL0157-1G TCIAL0157-5G
D-叔亮氨酸 ≥98.0% (by titrimetric analysis)
D-叔亮氨酸
分子式: C₆H₁₃NO₂
分子量: 131.2 g/mol
MDL编号: MFCD00004265
CAS号: 26782-71-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a49b3f75c1c66feb3f8dc1bbb485262cd866d922&vid=a49b3f75c1c66feb3f8dc1bbb485262cd866d922&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Optical purity (LC) min. 98.0 ee%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR