甲氨基乙醛缩二甲醇 ≥98.0% (by GC, titration analysis)
供应商: TCI
|
Warning
|
Synonyms:
N-甲基氨基乙醛二甲基乙缩醛, 甲氨基乙醛缩二甲醛
M0884-25ML
M0884-500ML
TCIAM0884-25MLEA
0
CNY
TCIAM0884-25ML
TCIAM0884-500ML
甲氨基乙醛缩二甲醇 ≥98.0% (by GC, titration analysis)
甲氨基乙醛缩二甲醇
分子式:
C₅H₁₃NO₂ 分子量: 119.16 g/mol 沸点: 140°C 闪点: 29°C |
MDL编号:
MFCD00008485 CAS号: 122-07-6 EINECS: 204-520-0 UN号: 1993 ADR: 3 III |
|
产品技术规格
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...