2,8-喹啉二醇 ≥98.0% (by GC, titration analysis)

供应商: TCI
Warning

Synonyms: 2,8-二羟基喹啉

D3819-25G D3819-5G
TCIAD3819-25GEA 0 CNY
TCIAD3819-25G TCIAD3819-5G
2,8-喹啉二醇 ≥98.0% (by GC, titration analysis)
2,8-喹啉二醇
分子式: C₉H₇NO₂
分子量: 161.16 g/mol
溶点: 290°C
存储条件: Ambient
MDL编号: MFCD00216696
CAS号: 15450-76-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=50bc82510bd84caaf80a260d8689d55846b57fea&vid=c5c317cfa639934030cf925d3b0b6829da222a91&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR