5-氯-8-喹啉醇 ≥98.0% (by GC, titration analysis)
供应商: TCI
|
Warning
|
Synonyms:
5-氯-8-羟基喹啉
C0645-25G
C0645-250G
TCIAC0645-25GEA
0
CNY
TCIAC0645-25G
TCIAC0645-250G
5-氯-8-喹啉醇 ≥98.0% (by GC, titration analysis)
5-氯-8-喹啉醇
分子式:
C₉H₆ClNO 分子量: 179.61 g/mol 沸点: 349 溶点: 125 存储条件: Ambient |
MDL编号:
MFCD00006788 CAS号: 130-16-5 EINECS: 204-978-1 Merck Index: 14,02422 |
|
产品技术规格
Appearance | White to Light yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 124.0 to 127.0 deg-C |
Solubility in hot Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...