2-喹啉甲醛 ≥97.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
Quinoline-2-carboxaldehyde, Quinoline-2-carbaldehyde, 2-Quinolinecarboxaldehyde
Q0064-1G
Q0064-5G
TCIAQ0064-1GEA
0
CNY
TCIAQ0064-1G
TCIAQ0064-5G
2-喹啉甲醛 ≥97.0% (by GC)
2-喹啉甲醛
分子式:
C₁₀H₇NO 分子量: 157.17 g/mol |
MDL编号:
MFCD00075032 CAS号: 5470-96-2 EINECS: 226-804-3 |
|
产品技术规格
Appearance | Light yellow to Brown powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 68.0 to 71.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...