1-(三甲基硅氧基)环己烯 ≥98.0% (by GC)

供应商: TCI
Warning

Synonyms: 1-Cyclohexenyloxytrimethylsilane, 1-环己烯氧基三甲基硅烷

T1556-10ML T1556-25ML
TCIAT1556-10MLEA 0 CNY
TCIAT1556-10ML TCIAT1556-25ML
1-(三甲基硅氧基)环己烯 ≥98.0% (by GC)
1-(三甲基硅氧基)环己烯
分子式: C₉H₁₈OSi
沸点: 165°C
闪点: 41°C
MDL编号: MFCD00001541
CAS号: 6651-36-1
EINECS: 229-675-1
UN号: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ce8e7d1141a44d0a53ddb8f07d31173211bf4a1e&vid=ce8e7d1141a44d0a53ddb8f07d31173211bf4a1e&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR