3,4-二甲氧基甲苯 ≥99.0% (通过GC)

供应商: TCI
Warning

D1986-25G D1986-500G
TCIAD1986-25GEA 0 CNY
TCIAD1986-25G TCIAD1986-500G
3,4-二甲氧基甲苯 ≥99.0% (通过GC)
3,4-二甲氧基甲苯
分子式: C₉H₁₂O₂
存储条件: Ambient
MDL编号: MFCD00016651
CAS号: 494-99-5
EINECS: 207-796-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c650d59d5da106de828fd131427b8433f131debc&vid=d96dafe4b2e3b4890d28d842a3527e50833a3fc3&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR